Assay Design



Gene Transcripts

Gene Symbol
Igf2bp1
Entrez Gene ID
140486
Gene Name
insulin-like growth factor 2 mRNA binding protein 1
Gene Aliases
AL024068, AW549074, CRD-BP, Crdbp, D030026A21Rik, D11Moh40e, D11Moh45, IMP-1, Neilsen, ZBP-1, Zbp1, mir-3063
Chromosome Location
Chr.11: 95957163 - 96005944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009951.4 NP_034081.1 15 - 15 7097 109
GenBank mRNA AK050850.1 - 1 - 1 1862 109
AK083554.1 - 1 - 1 108
BC052759.1 - 1 - 1 813 109

View Full Product Details