Assay Design
Gene Transcripts
- Gene Symbol
- Frs2
- Entrez Gene ID
-
327826
- Gene Name
- fibroblast growth factor receptor substrate 2
- Gene Aliases
-
4732458E18, C330018A15Rik, Frs2alpha, SNT1
- Chromosome Location
-
Chr.10: 117070127 - 117148474 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK049265.1 |
- |
- |
2131 |
68 |
View Full Product Details