Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177034.3 | NP_796008.2 | 12 - 13 | 2746 | 57 |
GenBank mRNA | AK032261.1 | - | 11 - 12 | 1510 | 57 |
AK147583.1 | - | 12 - 13 | 2736 | 57 | |
AK147623.1 | - | 12 - 13 | 2698 | 57 | |
BC141181.1 | - | 12 - 13 | 2564 | 57 |