Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008421.3 | NP_032447.1 | 2 - 2 | 7475 | 64 |
GenBank mRNA | AK082119.1 | - | 1 - 1 | 1555 | 64 |
AK086765.1 | - | 1 - 1 | 65 | ||
AK147476.1 | - | 2 - 2 | 7446 | 64 | |
AK147682.1 | - | 2 - 2 | 7444 | 64 | |
BC100546.1 | - | 1 - 1 | 1316 | 64 |