Assay Design
Gene Transcripts
- Gene Symbol
- Tra2b
- Entrez Gene ID
-
20462
- Gene Name
- transformer 2 beta homolog (Drosophila)
- Gene Aliases
-
5730405G21Rik, D16Ertd266e, SIG-41, Sfrs10, Silg41, TRA2beta
- Chromosome Location
-
Chr.16: 22245741 - 22266000 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK077418.1 |
- |
1 - 2 |
189 |
62 |
AK080378.1 |
- |
1 - 2 |
181 |
62 |
AK169483.1 |
- |
1 - 2 |
181 |
62 |
BC057996.1 |
- |
1 - 2 |
91 |
62 |
View Full Product Details