Assay Design
Gene Transcripts
- Gene Symbol
- Prdx6b
- Entrez Gene ID
-
320769
- Gene Name
- peroxiredoxin 6B
- Gene Aliases
-
1-cysPrx-P1, 4930414C22Rik, Aop2-rs1, Prdx5-rs1, Prdx6-ps1, Prdx6-rs1, Prdx6-rs1-ps
- Chromosome Location
-
Chr.2: 80292470 - 80295358 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_177256.5 |
NP_796230.1 |
1 - 1 |
1351 |
130 |
GenBank mRNA |
AK077012.1 |
- |
1 - 1 |
1347 |
130 |
View Full Product Details