Assay Design



Gene Transcripts

Gene Symbol
Umodl1
Entrez Gene ID
52020
Gene Name
uromodulin-like 1
Gene Aliases
D17Ertd488e
Chromosome Location
Chr.17: 30954640 - 31010710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177465.4 NP_803416.2 13 - 14 2638 57
GenBank mRNA AK040446.1 - 5 - 6 1139 57
AY771616.1 - 14 - 15 2555 57
AY771617.1 - 13 - 14 2642 57
AY771618.1 - 13 - 14 2297 57
BC051427.1 - 13 - 14 2293 57

View Full Product Details