Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177471.4 | NP_803422.2 | 2 - 3 | 210 | 86 |
GenBank mRNA | AK008760.1 | - | 2 - 3 | 181 | 86 |
AK079810.1 | - | 2 - 3 | 211 | 86 | |
AK135942.1 | - | 2 - 3 | 187 | 86 | |
AK170686.1 | - | 2 - 3 | 160 | 86 | |
BC096763.1 | - | 2 - 3 | 168 | 86 |