Assay Design



Gene Transcripts

Gene Symbol
Gdpd4
Entrez Gene ID
233537
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases
4921513O04
Chromosome Location
Chr.7: 97919944 - 98049663 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177696.3 NP_808364.2 3 - 4 387 68
GenBank mRNA AK029527.1 - 3 - 4 391 68
AK161500.1 - 3 - 4 354 68
BC140990.1 - 1 - 2 115 68

View Full Product Details