Assay Design



Gene Transcripts

Gene Symbol
Stac3
Entrez Gene ID
237611
Gene Name
SH3 and cysteine rich domain 3
Gene Aliases
9830125E18
Chromosome Location
Chr.10: 127501617 - 127508823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177707.3 NP_808375.1 9 - 10 1166 86
GenBank mRNA AK036516.1 - 9 - 10 1167 86
BC067208.1 - 9 - 10 946 86

View Full Product Details