Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177709.3 | NP_808377.1 | 2 - 3 | 796 | 59 |
GenBank mRNA | AK048522.1 | - | 2 - 3 | 796 | 59 |
AK158395.1 | - | 2 - 3 | 759 | 59 | |
BC119801.1 | - | 2 - 3 | 592 | 59 | |
BC122875.1 | - | 2 - 3 | 592 | 59 |