Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177794.3 | NP_808462.2 | 3 - 4 | 538 | 83 |
GenBank mRNA | AK028283.1 | - | 3 - 4 | 539 | 83 |
AK084351.1 | - | 3 - 4 | 516 | 83 | |
AK143866.1 | - | 3 - 4 | 516 | 83 | |
BC117502.1 | - | 4 - 5 | 542 | 83 | |
BC117503.1 | - | 3 - 4 | 460 | 83 |