Assay Design



Gene Transcripts

Gene Symbol
Wdr72
Entrez Gene ID
546144
Gene Name
WD repeat domain 72
Gene Aliases
AW558070, D230040E23
Chromosome Location
Chr.9: 74110331 - 74283203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033500.3 NP_001028672.2 3 - 4 657 67
GenBank mRNA AK052055.1 - 3 - 4 661 67
AK142559.1 - 3 - 4 637 67

View Full Product Details