Assay Design



Gene Transcripts

Gene Symbol
Klhl8
Entrez Gene ID
246293
Gene Name
kelch-like 8
Gene Aliases
2310001P09Rik, D5Ertd431e
Chromosome Location
Chr.5: 103862050 - 103911273 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178741.3 NP_848856.1 3 - 4 1048 61
GenBank mRNA AK031028.1 - 3 - 4 1049 61
AK034618.1 - 3 - 4 1014 61
AK049960.1 - 2 - 3 936 61
AY101769.1 - 2 - 3 793 61
BC086802.1 - 2 - 3 909 61

View Full Product Details