Assay Design
Gene Transcripts
- Gene Symbol
- Tyw1
- Entrez Gene ID
-
100929
- Gene Name
- tRNA-yW synthesizing protein 1 homolog (S. cerevisiae)
- Gene Aliases
-
AW125391, Rsafd1
- Chromosome Location
-
Chr.5: 130255572 - 130341567 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_178897.4 |
NP_849228.1 |
1 - 2 |
29 |
54 |
GenBank mRNA |
AK045279.1 |
- |
1 - 2 |
30 |
54 |
View Full Product Details