Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288586.1 | NP_001275515.1 | 1 - 2 | 204 | 106 |
NM_010786.4 | NP_034916.1 | 1 - 2 | 204 | 106 | |
GenBank mRNA | AK004719.1 | - | 1 - 2 | 163 | 106 |
AK004881.1 | - | 1 - 2 | 131 | 106 | |
AK088638.1 | - | 1 - 2 | 134 | 106 | |
AK168690.1 | - | 1 - 2 | 158 | 106 | |
BC050902.1 | - | 1 - 2 | 120 | 106 | |
BC092270.1 | - | 1 - 2 | 157 | 106 | |
BY323121.1 | - | 1 - 2 | 205 | 106 | |
CN703560.1 | - | 1 - 2 | 146 | 106 | |
X58876.1 | - | 1 - 2 | 112 | 106 |