Assay Design
Gene Transcripts
- Gene Symbol
- Hsd3b2
- Entrez Gene ID
-
15493
- Gene Name
- hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 98711107 - 98724594 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC040397.1 |
- |
- |
73 |
106 |
View Full Product Details