Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166024.1 | NP_001159496.1 | 11 - 12 | 1199 | 104 |
NM_023854.2 | NP_076343.2 | 10 - 11 | 1157 | 104 | |
GenBank mRNA | AF229439.1 | - | 10 - 11 | 966 | 104 |
AK009565.1 | - | 11 - 12 | 1023 | 104 | |
AK149837.1 | - | 10 - 11 | 992 | 104 | |
AK168489.1 | - | 10 - 11 | 967 | 104 | |
AK169144.1 | - | 10 - 11 | 980 | 104 | |
BC005495.1 | - | 10 - 11 | 1005 | 104 |