Assay Design



Gene Transcripts

Gene Symbol
Olfr5
Entrez Gene ID
18349
Gene Name
olfactory receptor 5
Gene Aliases
MOR103-8, ORL120
Chromosome Location
Chr.7: 6480216 - 6486909 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146914.2 NP_667125.1 2 - 2 451 160
GenBank mRNA BC104074.1 - 1 - 1 336 160
BC104075.1 - 1 - 1 346 160

View Full Product Details