Assay Design



Gene Transcripts

Gene Symbol
Rhox6
Entrez Gene ID
19202
Gene Name
reproductive homeobox 6
Gene Aliases
AV213605, AW548842, Psx1
Chromosome Location
Chr.X: 37827055 - 37829857 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008955.1 NP_032981.1 2 - 3 583 98
GenBank mRNA AF017453.1 - 2 - 3 583 98
AK076186.1 - 2 - 3 592 98
BC099477.1 - 2 - 3 579 98
DQ058643.1 - 2 - 3 583 98

View Full Product Details