Assay Design



Gene Transcripts

Gene Symbol
Zic2
Entrez Gene ID
22772
Gene Name
zinc finger protein of the cerebellum 2
Gene Aliases
HPE5, Ku
Chromosome Location
Chr.14: 122475384 - 122480328 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009574.3 NP_033600.3 1 - 2 1359 75
GenBank mRNA BE988982.1 - 1 - 2 389 75
D70848.2 - 1 - 2 1312 75

View Full Product Details