Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146292.1 | NP_001139764.1 | 3 - 4 | 803 | 141 |
NM_133195.3 | NP_573458.2 | 3 - 4 | 803 | 141 | |
GenBank mRNA | AK048427.1 | - | 3 - 4 | 980 | 141 |
BC048405.1 | - | 3 - 4 | 803 | 141 |