Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026636.2 | NP_080912.2 | 6 - 7 | 803 | 89 |
GenBank mRNA | AK077402.1 | - | 6 - 7 | 789 | 89 |
AK147083.1 | - | 6 - 7 | 798 | 89 | |
AK183167.1 | - | 2 - 3 | 86 | 89 | |
BC005692.1 | - | 6 - 7 | 790 | 89 |