Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164224.1 | NP_001157696.1 | 5 - 6 | 379 | 57 |
NM_026877.2 | NP_081153.1 | 6 - 7 | 600 | 57 | |
NM_198223.2 | NP_937866.1 | 5 - 6 | 379 | 57 | |
GenBank mRNA | AK088197.1 | - | 6 - 7 | 451 | 57 |
AK146061.1 | - | 5 - 6 | 380 | 57 | |
AY349132.1 | - | 4 - 5 | 369 | 57 | |
AY349133.1 | - | 6 - 7 | 517 | 57 | |
BC019177.1 | - | 6 - 7 | 543 | 57 | |
BC022115.1 | - | 6 - 7 | 547 | 57 | |
BC031153.1 | - | 6 - 7 | 532 | 57 |