Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028292.2 | NP_082568.1 | 10 - 11 | 1032 | 120 |
GenBank mRNA | AK028261.1 | - | 10 - 11 | 1067 | 120 |
AK076992.1 | - | 10 - 11 | 1443 | 120 | |
AK146051.1 | - | 10 - 11 | 1014 | 120 | |
AK154876.1 | - | 10 - 11 | 1105 | 120 | |
BC014867.1 | - | 10 - 11 | 1032 | 120 | |
BC029064.1 | - | 10 - 11 | 1020 | 120 |