Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028292.2 | NP_082568.1 | 12 - 13 | 1148 | 64 |
GenBank mRNA | AK028261.1 | - | 12 - 13 | 1183 | 64 |
AK146051.1 | - | 12 - 13 | 1130 | 64 | |
AK146268.1 | - | 12 - 13 | 1160 | 64 | |
AK154876.1 | - | 12 - 13 | 1221 | 64 | |
BC014867.1 | - | 12 - 13 | 1148 | 64 | |
BC029064.1 | - | 12 - 13 | 1136 | 64 |