Assay Design



Gene Transcripts

Gene Symbol
Nxf7
Entrez Gene ID
170722
Gene Name
nuclear RNA export factor 7
Gene Aliases
Nx7, Nxf6, nfx-a1, nxf-a, nxf-a2
Chromosome Location
Chr.X: 135579555 - 135598777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130888.1 NP_570958.1 5 - 6 463 53
GenBank mRNA AB178901.1 - 6 - 7 602 53
AJ305317.1 - 2 - 3 132 53
AJ305318.1 - 2 - 3 132 53
AY260550.1 - 5 - 6 463 53

View Full Product Details