Assay Design



Gene Transcripts

Gene Symbol
Zcchc9
Entrez Gene ID
69085
Gene Name
zinc finger, CCHC domain containing 9
Gene Aliases
1810019C21Rik
Chromosome Location
Chr.13: 91796520 - 91807997 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145453.2 NP_663428.2 3 - 4 649 72
GenBank mRNA AK044802.1 - 3 - 4 511 72
BC010687.1 - 2 - 3 564 72
BC024496.1 - 2 - 3 573 72

View Full Product Details