Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145567.1 | NP_663542.1 | 2 - 3 | 306 | 137 |
GenBank mRNA | AB051067.1 | - | 2 - 3 | 367 | 137 |
AK078175.1 | - | 2 - 3 | 257 | 137 | |
BC003914.1 | - | 2 - 3 | 306 | 137 |