Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178617.4 | NP_848732.3 | 8 - 9 | 910 | 117 |
GenBank mRNA | AK039240.1 | - | 8 - 9 | 726 | 117 |
AK050307.1 | - | 9 - 10 | 807 | 117 | |
AK081951.1 | - | 8 - 9 | 802 | 117 | |
AY278200.2 | - | 8 - 9 | 893 | 117 | |
BC067055.1 | - | 8 - 9 | 910 | 117 |