Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198168.3 | NP_937811.1 | 11 - 12 | 1710 | 82 |
GenBank mRNA | AK034167.1 | - | 11 - 12 | 1729 | 82 |
AK155872.1 | - | 11 - 12 | 1703 | 82 | |
AK171041.1 | - | 11 - 12 | 1681 | 82 | |
AK171178.1 | - | 11 - 12 | 1711 | 82 | |
AK171430.1 | - | 11 - 12 | 1718 | 82 | |
BC026670.1 | - | 7 - 8 | 566 | 82 | |
BC058977.1 | - | 11 - 12 | 1501 | 82 |