Assay Design



Gene Transcripts

Gene Symbol
Gorasp1
Entrez Gene ID
74498
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
5430411C10Rik, GOLPH5, GRASP65, P65
Chromosome Location
Chr.9: 119925673 - 119937558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028976.2 NP_083252.1 3 - 4 382 86
GenBank mRNA AK154986.1 - 3 - 4 359 86
AK170627.1 - 3 - 4 346 86
BC012251.1 - 3 - 4 382 86

View Full Product Details