Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030201.3 | NP_084477.1 | 2 - 3 | 418 | 67 |
NR_027492.1 | 2 - 3 | 418 | 67 | ||
GenBank mRNA | AK021006.1 | - | 2 - 3 | 390 | 67 |
AK030553.1 | - | 2 - 3 | 391 | 67 | |
AK034643.1 | - | 2 - 3 | 398 | 67 | |
AK049011.1 | - | 2 - 3 | 386 | 67 | |
AK167839.1 | - | 2 - 3 | 361 | 67 | |
BC039701.1 | - | 1 - 2 | 359 | 67 | |
BC085181.1 | - | 2 - 3 | 378 | 67 | |
BC094508.1 | - | 2 - 3 | 361 | 67 |