Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030201.3 | NP_084477.1 | 3 - 4 | 636 | 96 |
NR_027492.1 | 3 - 4 | 636 | 96 | ||
GenBank mRNA | AK030553.1 | - | 3 - 4 | 609 | 96 |
AK034643.1 | - | 3 - 4 | 616 | 96 | |
BC039701.1 | - | 2 - 3 | 577 | 96 | |
BC085181.1 | - | 3 - 4 | 596 | 96 | |
BC094508.1 | - | 3 - 4 | 579 | 96 |