Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173867.5 | NP_776292.1 | 4 - 5 | 706 | 57 |
GenBank mRNA | AK076040.1 | - | 4 - 5 | 637 | 57 |
AK076046.1 | - | - | 937 | 57 | |
AK129370.1 | - | 3 - 4 | 410 | 57 | |
BC020385.1 | - | 2 - 3 | 239 | 57 | |
BC060624.1 | - | 4 - 5 | 660 | 57 | |
BC086666.1 | - | 4 - 5 | 629 | 57 |