Assay Design



Gene Transcripts

Gene Symbol
Alkbh2
Entrez Gene ID
231642
Gene Name
alkB, alkylation repair homolog 2 (E. coli)
Gene Aliases
9530023G02, AU016977, Abh2, mABH2
Chromosome Location
Chr.5: 114123926 - 114130546 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175016.2 NP_778181.2 2 - 3 544 64
GenBank mRNA AK079195.1 - 2 - 3 381 64
BC062188.1 - 2 - 3 544 64

View Full Product Details