Assay Design



Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
108958
Gene Name
family with sequence similarity 73, member B
Gene Aliases
5730472N09Rik, AA408683, AI790341, C9orf54, R74766
Chromosome Location
Chr.2: 30364198 - 30385520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242407.1 NP_001229336.1 3 - 4 524 102
NM_175392.3 NP_780601.1 3 - 4 692 102
GenBank mRNA AK077618.1 - 3 - 4 485 102
AK140122.1 - 3 - 4 510 102
AK154200.1 - 3 - 4 513 102
AK165996.1 - 3 - 4 515 102
AK218176.1 - 1 - 2 55 102
BC051404.1 - 3 - 4 692 102

View Full Product Details