Assay Design



Gene Transcripts

Gene Symbol
Kif6
Entrez Gene ID
319991
Gene Name
kinesin family member 6
Gene Aliases
D130004B10Rik, D130084M03Rik
Chromosome Location
Chr.17: 49615104 - 49909847 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177052.3 NP_796026.2 5 - 6 571 63
GenBank mRNA AB001434.1 - 2 - 3 230 63
AK043404.1 - 5 - 6 528 63
AK083760.1 - 5 - 6 608 63

View Full Product Details