Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177771.5 | NP_808439.2 | 5 - 6 | 861 | 67 |
GenBank mRNA | AK044774.1 | - | 5 - 6 | 972 | 67 |
AK173029.1 | - | 5 - 6 | 861 | 67 | |
AK198584.1 | - | 2 - 3 | 213 | 67 | |
BC025563.1 | - | 5 - 6 | 857 | 67 | |
BC065167.1 | - | 4 - 5 | 558 | 67 | |
CB247282.1 | - | 2 - 3 | 209 | 67 | |
CK619719.1 | - | 3 - 4 | 467 | 67 |