Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178627.3 | NP_848742.1 | 7 - 8 | 1103 | 81 |
GenBank mRNA | AK081139.1 | - | 7 - 8 | 1063 | 81 |
AK083924.1 | - | 7 - 8 | 1104 | 81 | |
AK087446.1 | - | 7 - 8 | 1112 | 81 | |
AK088088.1 | - | 3 - 4 | 325 | 81 | |
AK150130.1 | - | 6 - 7 | 994 | 81 | |
AK152060.1 | - | 7 - 8 | 1144 | 81 | |
AK167552.1 | - | 7 - 8 | 1047 | 81 | |
AK220221.1 | - | 7 - 8 | 1119 | 81 | |
BC037652.1 | - | 7 - 8 | 1088 | 81 | |
BC046505.1 | - | 7 - 8 | 1032 | 81 | |
BC058225.1 | - | 7 - 8 | 1099 | 81 |