Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178628.5 | NP_848743.1 | 6 - 7 | 970 | 66 |
GenBank mRNA | AK046919.1 | - | 5 - 6 | 700 | 66 |
AK050339.1 | - | 6 - 7 | 807 | 66 | |
AK140185.1 | - | 5 - 6 | 714 | 66 | |
AK147311.1 | - | 5 - 6 | 4008 | 66 | |
BC050784.1 | - | 5 - 6 | 699 | 66 | |
BC057068.1 | - | 1 - 2 | 531 | 66 |