Assay Design



Gene Transcripts

Gene Symbol
Ano1
Entrez Gene ID
101772
Gene Name
anoctamin 1, calcium activated chloride channel
Gene Aliases
Tmem16a
Chromosome Location
Chr.7: 144588549 - 144752039 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242349.1 NP_001229278.1 7 - 8 1074 66
NM_178642.5 NP_848757.4 7 - 8 1071 66
GenBank mRNA AK032793.1 - 2 - 3 82 66
AK052589.1 - 7 - 8 1023 66
BC062959.1 - 7 - 8 960 66

View Full Product Details