Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170691.1 | NP_001164162.1 | 2 - 3 | 277 | 113 |
NM_178736.5 | NP_848851.3 | 2 - 3 | 282 | 113 | |
GenBank mRNA | AK029044.1 | - | 2 - 3 | 253 | 113 |
AK036607.1 | - | 2 - 3 | 260 | 113 | |
AK036746.1 | - | 2 - 3 | 222 | 113 | |
AK041137.1 | - | 2 - 3 | 183 | 113 | |
AK084950.1 | - | 2 - 3 | 282 | 113 |