Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181517.3 | NP_852658.2 | 16 - 17 | 1885 | 131 |
GenBank mRNA | AK039534.1 | - | 16 - 17 | 2037 | 131 |
AK079903.1 | - | 15 - 16 | 3395 | 131 | |
AY316153.1 | - | 16 - 17 | 1885 | 131 | |
BC053524.1 | - | 13 - 14 | 1568 | 131 | |
BC064825.1 | - | 7 - 8 | 841 | 131 |