Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181549.3 | NP_853527.2 | 3 - 4 | 537 | 108 |
GenBank mRNA | AK132888.1 | - | 5 - 6 | 1344 | 108 |
AY223868.1 | - | - | 745 | 108 | |
AY223869.1 | - | - | 716 | 108 | |
AY223870.1 | - | - | 720 | 108 | |
AY223871.1 | - | - | 626 | 108 | |
AY223872.1 | - | - | 691 | 108 | |
AY223873.1 | - | - | 597 | 108 |