Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019435.4 | NP_062308.2 | 3 - 3 | 769 | 70 |
GenBank mRNA | AK028121.1 | - | 3 - 3 | 490 | 70 |
BC027265.1 | - | 3 - 3 | 762 | 70 | |
BC096047.1 | - | 3 - 3 | 465 | 70 | |
Y08702.1 | - | 3 - 3 | 462 | 70 |