Assay Design
Gene Transcripts
- Gene Symbol
- Akirin1
- Entrez Gene ID
-
68050
- Gene Name
- akirin 1
- Gene Aliases
-
6330407G11Rik
- Chromosome Location
-
Chr.4: 123735195 - 123750299 on Build GRCm38
- Gene Symbol
- Appbp2
- Entrez Gene ID
-
66884
- Gene Name
- amyloid beta precursor protein (cytoplasmic tail) binding protein 2
- Gene Aliases
-
1300003O07Rik, AI465480, PAT1
- Chromosome Location
-
Chr.11: 85191308 - 85235120 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK153962.1 |
- |
1 - 1 |
|
98 |
View Full Product Details