Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025498.2 | NP_079774.1 | 4 - 4 | 539 | 69 |
GenBank mRNA | AK002440.1 | - | 4 - 4 | 487 | 69 |
AK006284.1 | - | 4 - 4 | 504 | 69 | |
AK007621.1 | - | 4 - 4 | 502 | 69 | |
BC024347.1 | - | 4 - 4 | 539 | 69 | |
BC081443.1 | - | 4 - 4 | 541 | 69 |