Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130864.3 | NP_570934.1 | 12 - 12 | 1661 | 106 |
GenBank mRNA | AK143187.1 | - | 12 - 12 | 1496 | 106 |
AY273811.1 | - | 12 - 12 | 1661 | 106 | |
BC012400.1 | - | 12 - 12 | 1477 | 106 | |
BC026669.1 | - | 5 - 5 | 713 | 106 |