Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178005.4 | NP_821072.1 | 2 - 2 | 1163 | 71 |
GenBank mRNA | AK036155.1 | - | 2 - 2 | 1158 | 71 |
AK043173.1 | - | 2 - 2 | 1019 | 71 | |
AK043784.1 | - | 2 - 2 | 1097 | 71 | |
AK044705.1 | - | 2 - 2 | 1165 | 71 | |
AK046774.1 | - | 2 - 2 | 1164 | 71 | |
AK046818.1 | - | 2 - 2 | 1058 | 71 | |
AK049920.1 | - | 2 - 2 | 1058 | 71 | |
AK122278.1 | - | 2 - 2 | 1149 | 71 | |
AY182027.1 | - | 1 - 1 | 735 | 71 |